Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032144.2 | NP_115520.2 | 1 | 636 | ||
GenBank mRNA | AF309646.1 | - | 1 | 229 | ||
AL136727.1 | - | 1 | 637 | |||
BC120999.1 | - | 1 | 474 | 40116526 | ||
CR533469.1 | - | 1 | 182 |
Gene Symbol: WTH3DI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077637.1 | NP_001071105.1 | 1 | 620 | ||
GenBank mRNA | AB232598.1 | - | 1 | 182 | ||
DB074386.1 | - | 2 | 507 |