Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032211.6 | NP_115587.6 | 4, 5 | 821 | ||
XM_005270216.1 | XP_005270273.1 | 4, 5 | 794 | |||
XM_006718020.1 | XP_006718083.1 | 5, 6 | 984 | |||
GenBank mRNA | AF338441.1 | - | 3, 4 | 670 | ||
AF395336.1 | - | 3, 4 | 670 | |||
AK172781.1 | - | 4, 5 | 764 | |||
AY036093.1 | - | 3, 4 | 676 | |||
BC013153.1 | - | 4, 5 | 821 | 3872314 | ||
BC035470.1 | - | 4, 5 | 821 |