Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290020.1 | NP_001276949.1 | 19 | 3105 | ||
NM_001290021.1 | NP_001276950.1 | 12 | 1931 | |||
NM_001312689.1 | NP_001299618.1 | 14 | 2322 | |||
NM_018076.4 | NP_060546.2 | 19 | 3058 | |||
XM_011519527.1 | XP_011517829.1 | 19 | 3105 | |||
XM_011519528.1 | XP_011517830.1 | 19 | 3755 | |||
XM_011519533.2 | XP_011517835.1 | 14 | 2279 | |||
XM_011519534.2 | XP_011517836.1 | 14 | 2412 | |||
XM_011519535.2 | XP_011517837.1 | 13 | 2164 | |||
XM_011519537.2 | XP_011517839.1 | 10 | 1839 | |||
XM_017016368.1 | XP_016871857.1 | 19 | 3105 | |||
XM_017016369.1 | XP_016871858.1 | 19 | 3105 | |||
XM_017016370.1 | XP_016871859.1 | 18 | 2895 | |||
GenBank mRNA | AK001679.1 | - | 19 | 3058 | ||
AK292521.1 | - | 19 | 3061 | |||
AK302048.1 | - | 12 | 1885 | |||
AK302419.1 | - | 14 | 2654 | |||
AK302627.1 | - | 14 | 2322 | |||
AL136859.1 | - | 8 | 1444 | |||
BC140846.1 | - | 18 | 2986 | 9021714 |