Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032169.4 | NP_115545.3 | 11 | 1681 | ||
NR_132426.1 | 11 | 1681 | ||||
NR_132427.1 | 11 | 1681 | ||||
GenBank mRNA | AJ608287.1 | - | 11 | 1632 | ||
AK022654.1 | - | 4 | 275 | |||
AK131265.1 | - | 13 | 1478 | |||
AK298627.1 | - | 3 | 488 | |||
AK308380.1 | - | 12 | 1271 | |||
AL832873.1 | - | 11 | 1632 | |||
AL833721.1 | - | 11 | 1199 | |||
AX803783.1 | - | 11 | 1538 | |||
AX803785.1 | - | 11 | 1301 | |||
BC019607.1 | - | 6 | 577 | |||
BC125204.1 | - | 11 | 1616 | 40123454 |
Gene Symbol: NPHP3-ACAD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037804.1 | 37 | 5303 | |||
GenBank mRNA | AB082531.1 | - | 37 | 5303 |