Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032211.6 | NP_115587.6 | 8 | 1385 | ||
XM_005270216.1 | XP_005270273.1 | 8 | 1361 | |||
XM_006718020.1 | XP_006718083.1 | 9 | 1551 | |||
GenBank mRNA | AF338441.1 | - | 7 | 1234 | ||
AF395336.1 | - | 7 | 1234 | |||
AK172781.1 | - | 8 | 1328 | |||
AY036093.1 | - | 7 | 1240 | |||
BC007522.1 | - | 1 | 101 | |||
BC013153.1 | - | 8 | 1385 | 3872314 | ||
BC035470.1 | - | 8 | 1385 |