Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC37A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287498.1 | NP_001274427.1 | 9 | 933 | ||
NM_032295.3 | NP_115671.1 | 9 | 933 | |||
NM_207113.2 | NP_996996.1 | 9 | 933 | |||
XM_011516626.2 | XP_011514928.1 | 11 | 1430 | |||
XM_011516627.2 | XP_011514929.1 | 11 | 1429 | |||
XM_017012712.1 | XP_016868201.1 | 11 | 1406 | |||
XM_017012713.1 | XP_016868202.1 | 11 | 1425 | |||
XM_017012714.1 | XP_016868203.1 | 9 | 1203 | |||
XM_017012717.1 | XP_016868206.1 | 11 | 1425 | |||
GenBank mRNA | AK074823.1 | - | 9 | 894 | ||
AK299276.1 | - | 10 | 979 | |||
AK300265.1 | - | 8 | 1263 | |||
AL136583.1 | - | 9 | 840 | |||
BC028380.1 | - | 9 | 883 | 4811814 | ||
BC046567.1 | - | 9 | 885 | 5246993 |