Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323041.1 | NP_001309970.1 | 3, 4 | 773 | ||
NM_001323042.1 | NP_001309971.1 | 3, 4 | 257 | |||
NM_001323043.1 | NP_001309972.1 | 3, 4 | 658 | |||
NM_001323047.1 | NP_001309976.1 | 3, 4 | 773 | |||
NM_001323048.1 | NP_001309977.1 | 3, 4 | 658 | |||
NM_001323049.1 | NP_001309978.1 | 2, 3 | 947 | |||
NM_001323052.1 | NP_001309981.1 | 3, 4 | 658 | |||
NM_001323053.1 | NP_001309982.1 | 3, 4 | 773 | |||
NM_006608.2 | NP_006599.2 | 2, 3 | 562 | |||
NR_136565.1 | 3, 4 | 658 | ||||
NR_136566.1 | 3, 4 | 773 | ||||
NR_136567.1 | 3, 4 | 658 | ||||
GenBank mRNA | AJ011863.1 | - | 2, 3 | 141 | ||
AK294749.1 | - | 2, 3 | 288 | |||
BC002447.2 | - | 3, 4 | 706 | 3346990 | ||
CX763183.1 | - | 2, 3 | 438 |