Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286771.2 | NP_001273700.1 | 5 | 654 | ||
NM_015574.1 | NP_056389.1 | 5 | 1036 | |||
NM_032217.4 | NP_115593.3 | 5 | 1036 | |||
NM_198889.2 | NP_942592.1 | 5 | 1036 | |||
XM_005265671.3 | XP_005265728.1 | 5 | 1380 | |||
XM_005265672.3 | XP_005265729.1 | 5 | 1380 | |||
XM_005265673.3 | XP_005265730.1 | 5 | 1380 | |||
XM_017008011.1 | XP_016863500.1 | 5 | 611 | |||
XM_017008012.1 | XP_016863501.1 | 5 | 611 | |||
XM_017008013.1 | XP_016863502.1 | 5 | 611 | |||
XM_017008014.1 | XP_016863503.1 | 5 | 1380 | |||
XM_017008015.1 | XP_016863504.1 | 5 | 611 | |||
XM_017008016.1 | XP_016863505.1 | 5 | 1380 | |||
XM_017008017.1 | XP_016863506.1 | 5 | 611 | |||
GenBank mRNA | AF308285.1 | - | 5 | 1023 | ||
AK300302.1 | - | 5 | 610 | |||
AK307581.1 | - | 5 | 1036 | |||
AL833287.1 | - | 2 | 213 | |||
BC004173.2 | - | 5 | 1017 | |||
BC007747.2 | - | 3 | 366 | |||
BC043394.1 | - | 5 | 1005 | |||
BX648035.1 | - | 5 | 609 |