Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032323.2 | NP_115699.1 | 2 | 368 | ||
NR_026678.1 | 2 | 393 | ||||
GenBank mRNA | AK056816.1 | - | 2 | 333 | ||
AK315767.1 | - | 2 | 336 | |||
BC005094.1 | - | 2 | 358 | 3955668 | ||
DA678865.1 | - | 2 | 393 | |||
DA880152.1 | - | 2 | 368 |