Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032361.3 | NP_115737.1 | 1, 2 | 373 | ||
GenBank mRNA | BC006849.1 | - | 1, 2 | 335 | 3451612 | |
BC066325.1 | - | 1, 2 | 330 | 5303134 | ||
BC068499.1 | - | 1, 2 | 292 | 5262880 | ||
BG502664.1 | - | 1, 2 | 94 | |||
HY009859.1 | - | 1, 2 | 374 |
Gene Symbol: LOC728554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003615.2 | 1, 2 | 599 |