Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032472.3 | NP_115861.1 | 5 | 445 | ||
NM_130906.2 | NP_570981.1 | 5 | 433 | |||
XM_005246651.3 | XP_005246708.1 | 5 | 364 | |||
XM_005246652.4 | XP_005246709.1 | 5 | 352 | |||
XM_011511358.2 | XP_011509660.1 | 5 | 538 | |||
XM_017004352.1 | XP_016859841.1 | 5 | 555 | |||
XM_017004353.1 | XP_016859842.1 | 5 | 294 | |||
XM_017004354.1 | XP_016859843.1 | 5 | 728 | |||
XM_017004355.1 | XP_016859844.1 | 5 | 752 | |||
XM_017004356.1 | XP_016859845.1 | 5 | 716 | |||
XM_017004357.1 | XP_016859846.1 | 6 | 635 | |||
XM_017004358.1 | XP_016859847.1 | 6 | 458 | |||
XM_017004359.1 | XP_016859848.1 | 6 | 642 | |||
XM_017004360.1 | XP_016859849.1 | 4 | 455 | |||
GenBank mRNA | AF251049.1 | - | 5 | 445 | ||
AF271652.1 | - | 5 | 368 | |||
AF414127.1 | - | 4 | 369 | |||
AK297804.1 | - | 5 | 364 | |||
AY257468.1 | - | 4 | 489 | |||
BC007693.2 | - | 5 | 874 | 3956681 | ||
CB961908.1 | - | 5 | 488 | |||
KU178646.1 | - | 3 | 192 | |||
KU178647.1 | - | 4 | 245 | |||
KU178648.1 | - | 3 | 204 |