Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102559.1 | NP_001096029.1 | 3 | 269 | ||
NM_001102560.1 | NP_001096030.1 | 3 | 269 | |||
NM_032483.3 | NP_115872.2 | 3 | 269 | |||
XM_005273661.4 | XP_005273718.2 | 3 | 463 | |||
XM_005273663.3 | XP_005273720.2 | 3 | 476 | |||
XM_011544670.2 | XP_011542972.1 | 3 | 484 | |||
XM_011544671.1 | XP_011542973.1 | 3 | 253 | |||
XM_011544672.1 | XP_011542974.1 | 3 | 609 | |||
XM_011544674.2 | XP_011542976.1 | 3 | 482 | |||
XM_011544675.2 | XP_011542977.1 | 3 | 480 | |||
XM_017013905.1 | XP_016869394.1 | 3 | 482 | |||
XM_017013906.1 | XP_016869395.1 | 3 | 482 | |||
XM_017013907.1 | XP_016869396.1 | 3 | 609 | |||
GenBank mRNA | AF212238.1 | - | 3 | 223 | ||
AK292118.1 | - | 3 | 194 | |||
BC033025.1 | - | 2 | 329 | 5267610 | ||
BC106014.1 | - | 3 | 239 | 3877263 | ||
BQ272061.1 | - | 3 | 249 | |||
DB458595.1 | - | 3 | 270 |