Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007144.2 | NP_009075.1 | 4, 5 | 460 | ||
XM_005257640.2 | XP_005257697.1 | 5, 6 | 919 | |||
XM_005257641.4 | XP_005257698.1 | 3, 4 | 1141 | |||
XM_005257642.3 | XP_005257699.1 | 3, 4 | 1538 | |||
XM_017025016.1 | XP_016880505.1 | 5, 6 | 1479 | |||
GenBank mRNA | BC004858.2 | - | 2, 3 | 231 | 3841545 | |
BC024255.1 | - | 4, 5 | 456 | 3605568 | ||
BX647429.1 | - | 5, 6 | 919 | |||
D13969.1 | - | 3, 4 | 418 |