Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128612.2 | NP_001122084.1 | 5 | 814 | ||
NM_001128613.2 | NP_001122085.1 | 4 | 829 | |||
NM_001271831.1 | NP_001258760.1 | 4 | 715 | |||
NM_032344.3 | NP_115720.1 | 5 | 928 | |||
GenBank mRNA | BC006129.1 | - | 5 | 912 | 3625545 | |
BM923060.1 | - | 4 | 714 |