Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032360.3 | NP_115736.1 | 1, 2 | 851 | ||
GenBank mRNA | AK314112.1 | - | 1, 2 | 826 | ||
AL537685.3 | - | 1, 2 | 849 | |||
AL561590.3 | - | 1, 2 | 846 | |||
AY606133.1 | - | 1, 2 | 264 | |||
BC006505.2 | - | 1, 2 | 308 | 2821736 |