Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102559.1 | NP_001096029.1 | 6 | 534 | ||
NM_032483.3 | NP_115872.2 | 6 | 534 | |||
XM_005273661.4 | XP_005273718.2 | 6 | 728 | |||
XM_011544671.1 | XP_011542973.1 | 6 | 518 | |||
XM_011544672.1 | XP_011542974.1 | 6 | 874 | |||
XM_017013907.1 | XP_016869396.1 | 6 | 874 | |||
GenBank mRNA | AF212238.1 | - | 6 | 488 | ||
AK292118.1 | - | 6 | 459 | |||
BC033025.1 | - | 5 | 594 | 5267610 | ||
BC106014.1 | - | 6 | 504 | 3877263 |