Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033002.3 | NP_001028174.2 | 6 | 1109 | ||
NM_001160243.1 | NP_001153715.1 | 6 | 1109 | |||
NR_027679.1 | 5 | 997 | ||||
NR_027682.1 | 5 | 1045 | ||||
NR_027683.1 | 5 | 997 | ||||
NR_027685.1 | 4 | 933 | ||||
GenBank mRNA | AK096001.1 | - | 5 | 453 | ||
AK295394.1 | - | 6 | 550 | |||
AK300409.1 | - | 5 | 718 | |||
AY680654.1 | - | 6 | 539 | |||
AY680656.1 | - | 5 | 475 | |||
AY680657.1 | - | 5 | 427 | |||
AY680659.1 | - | 4 | 363 | |||
AY775314.1 | - | 6 | 1109 | |||
AY775315.1 | - | 5 | 1045 | |||
AY775317.1 | - | 5 | 1262 | |||
AY775320.1 | - | 4 | 933 | |||
AY775323.1 | - | 5 | 997 | |||
BC004451.1 | - | 5 | 488 | 2820882 | ||
BC013831.1 | - | 6 | 580 | |||
BC051849.2 | - | 6 | 542 | 5493799 | ||
DA349558.1 | - | 4 | 372 |