Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPNS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032330.1 | NP_115706.1 | 1 | 187 | ||
GenBank mRNA | AY052551.1 | - | 1 | 159 | ||
BC005397.1 | - | 1 | 175 | 3685903 | ||
BC006000.1 | - | 1 | 187 | 4092675 |