Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098632.2 | NP_001092102.1 | 5 | 1548 | ||
NM_001098633.3 | NP_001092103.1 | 5 | 1336 | |||
NM_001278159.1 | NP_001265088.1 | 5 | 1561 | |||
NM_001278160.1 | NP_001265089.1 | 5 | 1372 | |||
NM_032375.5 | NP_115751.3 | 5 | 1970 | |||
GenBank mRNA | AK055511.1 | - | 5 | 1561 | ||
AK123687.1 | - | 5 | 1935 | |||
BC007416.2 | - | 5 | 1856 | 2988648 | ||
BC015562.1 | - | 6 | 1140 | |||
BC051844.2 | - | 5 | 1440 | 6164197 |