Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032413.3 | NP_115789.1 | 4 | 681 | ||
NM_197955.2 | NP_922946.1 | 5 | 567 | |||
GenBank mRNA | AB026707.1 | - | 5 | 461 | ||
AF228422.1 | - | 4 | 602 | |||
AK026298.1 | - | 4 | 599 | |||
BC021173.2 | - | 5 | 487 | 5269504 |