Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012188.4 | NP_036320.2 | 2 | 1101 | ||
NM_144769.2 | NP_658982.1 | 2 | 816 | |||
GenBank mRNA | AK292176.1 | - | 2 | 1117 | ||
AY707089.1 | - | 2 | 690 | |||
BC029778.1 | - | 2 | 816 | 5185923 | ||
L13203.1 | - | 2 | 1053 |