Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252675.1 | NP_001239604.1 | 1 | 404 | ||
NM_001252677.1 | NP_001239606.1 | 1 | 404 | |||
NM_032501.3 | NP_115890.2 | 1 | 404 | |||
XM_006723659.1 | XP_006723722.1 | 1 | 365 | |||
XM_011529388.1 | XP_011527690.1 | 1 | 365 | |||
GenBank mRNA | AK094414.1 | - | 1 | 1203 | ||
AK127566.1 | - | 1 | 350 | |||
AK314440.1 | - | 1 | 327 | |||
AL832939.1 | - | 1 | 88 | |||
BC039261.1 | - | 1 | 363 | 5298849 | ||
BC044588.1 | - | 1 | 329 | 5260496 | ||
BC055008.1 | - | 1 | 329 | 4838976 | ||
BC114612.1 | - | 1 | 195 | |||
BM150326.1 | - | 1 | 404 |