Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318124.1 | NP_001305053.1 | 1 | 226 | ||
NM_001318125.1 | NP_001305054.1 | 1 | 226 | |||
NM_032562.4 | NP_115951.2 | 1 | 226 | |||
XM_005270244.4 | XP_005270301.1 | 1 | 451 | |||
GenBank mRNA | AF339053.1 | - | 1 | 185 | ||
AF349540.1 | - | 1 | 129 | |||
AK311349.1 | - | 1 | 213 | |||
AK313712.1 | - | 1 | 189 | |||
AY358032.1 | - | 1 | 159 | |||
BC093996.1 | - | 1 | 148 | 7939841 | ||
BC111946.1 | - | 1 | 148 | 8327414 | ||
BC143532.1 | - | 1 | 148 | 9052041 | ||
BP274516.1 | - | 1 | 226 | |||
DA536331.1 | - | 1 | 213 |