Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168399.1 | NP_001161871.1 | 3 | 1021 | ||
NM_001168400.1 | NP_001161872.1 | 3 | 1018 | |||
NM_001168401.1 | NP_001161873.1 | 3 | 763 | |||
NM_032621.3 | NP_116010.1 | 3 | 835 | |||
GenBank mRNA | AF251053.1 | - | 3 | 667 | ||
AY833560.1 | - | 3 | 767 | |||
BC015522.1 | - | 3 | 713 | 3914401 |