Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001928.2 | NP_001001928.1 | 8 | 6732 | ||
NM_005036.4 | NP_005027.2 | 9 | 6816 | |||
GenBank mRNA | AB073605.1 | - | 1 | 709 | ||
AK091885.1 | - | 1 | 521 | |||
BC004162.2 | - | 1 | 2875 | |||
BC071932.1 | - | 1 | 562 |