Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAPS2 SIRNA Assays ›
Gene Symbol:
CAPS2
Gene Name:
calcyphosine 2
Gene Aliases:
UG0636c06
Chromosome Location:
Chr.12: 75275979 - 75390941 on Build GRCh38
UniGene ID:
Hs.733835
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
34523
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAPS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286548.1
NP_001273477.1
18
2242
NM_001286549.1
NP_001273478.1
11
1417
XM_005269192.1
XP_005269249.1
16
1793
XM_006719648.2
XP_006719711.2
18
2114
XM_006719649.2
XP_006719712.2
17
2018
XM_011538878.1
XP_011537180.1
18
2079
XM_011538886.2
XP_011537188.1
17
2048
XM_011538887.2
XP_011537189.1
16
1952
XM_011538888.1
XP_011537190.1
14
1633
XM_011538889.2
XP_011537191.1
18
1985
XM_011538892.1
XP_011537194.1
15
1679
XM_017020034.1
XP_016875523.1
18
1941
XM_017020035.1
XP_016875524.1
13
1537
XM_017020036.1
XP_016875525.1
19
2204
XM_017020037.1
XP_016875526.1
16
1833
XM_017020038.1
XP_016875527.1
14
1701
XM_017020039.1
XP_016875528.1
12
1401
XM_017020040.1
XP_016875529.1
7
973
XM_017020041.1
XP_016875530.1
16
1939
XM_017020042.1
XP_016875531.1
16
1967
XM_017020043.1
XP_016875532.1
12
1784
XM_017020044.1
XP_016875533.1
11
1378
XM_017020045.1
XP_016875534.1
12
1820
XM_017020046.1
XP_016875535.1
11
1377
XM_017020047.1
XP_016875536.1
10
4830
GenBank mRNA
AF251056.1
-
18
2242
AF351611.1
-
17
1823
AK092274.1
-
12
1330
BC131724.1
-
11
1416
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Process(es)
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0