Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321967.1 | NP_001308896.1 | 7 | 809 | ||
NM_001321969.1 | NP_001308898.1 | 8 | 899 | |||
NM_032810.3 | NP_116199.2 | 7 | 1181 | |||
NR_135914.1 | 7 | 809 | ||||
XM_005270252.4 | XP_005270309.1 | 7 | 903 | |||
XM_011540302.1 | XP_011538604.1 | 5 | 574 | |||
XM_011540303.2 | XP_011538605.1 | 7 | 856 | |||
XM_017016847.1 | XP_016872336.1 | 7 | 1050 | |||
XM_017016849.1 | XP_016872338.1 | 8 | 1022 | |||
XM_017016850.1 | XP_016872339.1 | 8 | 1073 | |||
GenBank mRNA | AF361493.1 | - | 6 | 1015 | ||
AK027506.1 | - | 4 | 409 | |||
AK075223.1 | - | 7 | 1110 | |||
AL834370.1 | - | 6 | 594 | |||
BC010868.1 | - | 3 | 244 | |||
BC063530.1 | - | 6 | 640 | |||
BC073998.1 | - | 6 | 752 | 6199813 |