Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSH2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032855.3 | NP_116244.1 | 4, 5 | 665 | ||
NR_111903.1 | 4, 5 | 665 | ||||
NR_111904.1 | 4, 5 | 665 | ||||
XM_011528386.2 | XP_011526688.1 | 2, 3 | 260 | |||
XM_011528387.2 | XP_011526689.1 | 2, 3 | 251 | |||
XM_011528389.2 | XP_011526691.1 | 2, 3 | 275 | |||
XM_011528390.1 | XP_011526692.1 | 2, 3 | 251 | |||
XM_017027402.1 | XP_016882891.1 | 2, 3 | 251 | |||
XM_017027403.1 | XP_016882892.1 | 2, 3 | 251 | |||
XM_017027404.1 | XP_016882893.1 | 2, 3 | 261 | |||
GenBank mRNA | AK027792.1 | - | 2, 3 | 245 | ||
AK298682.1 | - | 2, 3 | 208 | |||
AK307563.1 | - | 2, 3 | 208 | |||
AY319652.1 | - | 4, 5 | 665 | |||
BC025237.2 | - | 1, 2 | 165 | 4853087 |