Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032861.3 | NP_116250.3 | 4, 5 | 403 | ||
NR_073096.1 | 4, 5 | 403 | ||||
XM_006715586.2 | XP_006715649.1 | 3, 4 | 332 | |||
XM_011536196.2 | XP_011534498.1 | 3, 4 | 426 | |||
XM_011536198.2 | XP_011534500.1 | 2, 3 | 328 | |||
GenBank mRNA | AK027823.1 | - | 4, 5 | 393 | ||
AK125878.1 | - | 4, 5 | 1495 | |||
BC001705.1 | - | 4, 5 | 360 | 3350731 | ||
BC028594.1 | - | 5, 6 | 3602 | |||
BX648104.1 | - | 3, 4 | 335 | |||
DA232092.1 | - | 4, 5 | 403 |