Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318391.1 | NP_001305320.1 | 8 | 1001 | ||
NM_032830.2 | NP_116219.2 | 8 | 1055 | |||
XM_005256205.3 | XP_005256262.2 | 4 | 586 | |||
GenBank mRNA | AB075868.2 | - | 8 | 993 | ||
AK027419.1 | - | 5 | 610 | |||
AK027445.1 | - | 6 | 678 | |||
AK027634.1 | - | 8 | 1007 | |||
AK027675.1 | - | 8 | 1004 | |||
AK074795.1 | - | 8 | 1021 | |||
AK310104.1 | - | 8 | 957 | |||
BC000167.2 | - | 4 | 500 | |||
BC009348.2 | - | 8 | 987 | 4131625 | ||
BX647265.1 | - | 8 | 969 |