Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPND | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159846.2 | NP_001153318.1 | 11 | 1484 | ||
NM_001300862.1 | NP_001287791.1 | 13 | 1634 | |||
NM_032868.5 | NP_116257.2 | 12 | 1544 | |||
XM_006722926.2 | XP_006722989.1 | 13 | 1508 | |||
GenBank mRNA | AK027887.1 | - | 11 | 1457 | ||
BC032652.1 | - | 12 | 1501 | 5503607 |