Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286491.1 | NP_001273420.1 | 15 | 2046 | ||
NM_032892.4 | NP_116281.2 | 14 | 1727 | |||
NR_104455.1 | 15 | 2043 | ||||
GenBank mRNA | AK090572.1 | - | 14 | 1659 | ||
AK290013.1 | - | 3 | 521 | |||
BC043486.2 | - | 15 | 1793 | |||
BC053647.1 | - | 14 | 1664 | 5578205 |