Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201572.1 | NP_001188501.1 | 6 | 813 | ||
NM_004206.3 | NP_004197.1 | 6 | 875 | |||
NM_032970.3 | NP_116752.1 | 6 | 875 | |||
XM_005265533.1 | XP_005265590.1 | 7 | 1049 | |||
XM_005265534.1 | XP_005265591.1 | 7 | 949 | |||
XM_005265535.1 | XP_005265592.1 | 7 | 1007 | |||
XM_011534212.1 | XP_011532514.1 | 6 | 727 | |||
XM_017007438.1 | XP_016862927.1 | 7 | 1049 | |||
XM_017007439.1 | XP_016862928.1 | 7 | 1069 | |||
GenBank mRNA | AF039568.1 | - | 5 | 665 | ||
AK074576.1 | - | 6 | 874 | |||
AK292856.1 | - | 6 | 809 | |||
AK293133.1 | - | 7 | 925 | |||
BC006178.1 | - | 6 | 780 | 4054516 | ||
BC018437.1 | - | 6 | 783 | 3445481 | ||
BX377531.2 | - | 6 | 814 | |||
CR749670.1 | - | 4 | 429 | |||
KU178463.1 | - | 5 | 665 | |||
KU178464.1 | - | 5 | 665 | |||
KU178465.1 | - | 4 | 501 |