Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172648.3 | NP_001166119.1 | 13 | 2166 | ||
NM_001172649.3 | NP_001166120.1 | 12 | 2263 | |||
NM_001291106.1 | NP_001278035.1 | 13 | 2410 | |||
NM_001291107.1 | NP_001278036.1 | 13 | 2410 | |||
NM_007185.6 | NP_009116.3 | 13 | 2413 | |||
XM_006711139.2 | XP_006711202.1 | 14 | 1641 | |||
XM_006711141.2 | XP_006711204.1 | 14 | 1638 | |||
XM_011509111.2 | XP_011507413.1 | 12 | 2108 | |||
XM_011509112.2 | XP_011507414.1 | 12 | 1504 | |||
XM_011509114.2 | XP_011507416.1 | 12 | 1472 | |||
XM_017000145.1 | XP_016855634.1 | 12 | 1479 | |||
XM_017000146.1 | XP_016855635.1 | 12 | 1476 | |||
GenBank mRNA | AB209264.1 | - | 13 | 2166 | ||
AF284423.1 | - | 4 | 640 | |||
AK298852.1 | - | 7 | 1036 | |||
AY165003.1 | - | 13 | 2403 | |||
BC004145.2 | - | 9 | 789 | |||
BC041088.1 | - | 12 | 1495 | |||
BC052491.1 | - | 13 | 2384 | 4820737 | ||
BC068008.1 | - | 12 | 1440 | |||
U80746.1 | - | 10 | 1292 | |||
U80759.1 | - | 10 | 1299 |