Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171971.2 | NP_001165442.1 | 3, 4 | 433 | ||
NM_033100.3 | NP_149091.1 | 3, 4 | 433 | |||
XM_011540337.1 | XP_011538639.1 | 2, 3 | 924 | |||
XM_011540338.1 | XP_011538640.1 | 2, 3 | 924 | |||
XM_011540339.1 | XP_011538641.1 | 3, 4 | 307 | |||
XM_011540340.2 | XP_011538642.1 | 2, 3 | 924 | |||
GenBank mRNA | AB053448.1 | - | 3, 4 | 306 | ||
AB593154.1 | - | 3, 4 | 433 | |||
AB593155.1 | - | 3, 4 | 419 | |||
BC038799.1 | - | 3, 4 | 423 | 5492127 |