Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RERE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042681.1 | NP_001036146.1 | 12 | 1859 | ||
NM_001042682.1 | NP_001036147.1 | 2 | 158 | |||
NM_012102.3 | NP_036234.3 | 13 | 2044 | |||
XM_005263464.2 | XP_005263521.1 | 12 | 1798 | |||
XM_005263466.1 | XP_005263523.1 | 6 | 681 | |||
XM_011541510.1 | XP_011539812.1 | 11 | 1784 | |||
XM_011541511.1 | XP_011539813.1 | 12 | 1910 | |||
XM_017001358.1 | XP_016856847.1 | 12 | 1549 | |||
XM_017001359.1 | XP_016856848.1 | 12 | 2054 | |||
GenBank mRNA | AB007927.2 | - | 11 | 1194 | ||
AB036737.1 | - | 13 | 1870 | |||
AF016005.1 | - | 2 | 137 | |||
CD674939.1 | - | 2 | 158 |