Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135610.1 | NP_001129082.1 | 3 | 226 | ||
NM_012231.4 | NP_036363.2 | 3 | 956 | |||
NM_015866.4 | NP_056950.2 | 3 | 956 | |||
XM_017002253.1 | XP_016857742.1 | 2 | 178 | |||
XM_017002254.1 | XP_016857743.1 | 2 | 178 | |||
XM_017002255.1 | XP_016857744.1 | 2 | 188 | |||
XM_017002256.1 | XP_016857745.1 | 3 | 257 | |||
XM_017002257.1 | XP_016857746.1 | 2 | 186 | |||
XM_017002264.1 | XP_016857753.1 | 2 | 1517 | |||
GenBank mRNA | U17838.1 | - | 3 | 955 |