Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004342.6 | NP_004333.1 | 6 | 1167 | ||
NM_033138.3 | NP_149129.2 | 7 | 1932 | |||
NM_033139.3 | NP_149130.1 | 5 | 982 | |||
NM_033140.3 | NP_149131.1 | 4 | 904 | |||
NM_033157.3 | NP_149347.2 | 7 | 1245 | |||
XM_017012650.1 | XP_016868139.1 | 5 | 1467 | |||
XM_017012651.1 | XP_016868140.1 | 5 | 780 | |||
XM_017012652.1 | XP_016868141.1 | 5 | 941 | |||
XM_017012653.1 | XP_016868142.1 | 4 | 702 | |||
XM_017012654.1 | XP_016868143.1 | 4 | 904 | |||
GenBank mRNA | AF247820.3 | - | 6 | 930 | ||
AK289686.1 | - | 7 | 1238 | |||
AK298524.1 | - | 6 | 793 | |||
AK300282.1 | - | 4 | 873 | |||
AK304729.1 | - | 6 | 714 | |||
BC014035.2 | - | 4 | 867 | |||
BC040354.1 | - | 6 | 902 | 4753285 | ||
BX648808.1 | - | 4 | 865 | |||
D90452.1 | - | 5 | 772 | |||
D90453.1 | - | 4 | 694 | |||
M64110.1 | - | 5 | 812 | |||
M83216.1 | - | 7 | 1695 |