Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033198.3 | NP_149975.1 | 12 | 2337 | ||
GenBank mRNA | AF161427.1 | - | 6 | 1197 | ||
AK025825.1 | - | 5 | 1102 | |||
AK055512.1 | - | 7 | 1167 | |||
AK075428.1 | - | 12 | 1992 | |||
AK127162.1 | - | 3 | 1905 | |||
AK297046.1 | - | 11 | 1856 | |||
AY359112.1 | - | 11 | 2232 | |||
BC001319.2 | - | 4 | 1030 | |||
BC007301.1 | - | 10 | 1721 | |||
BC069228.1 | - | 12 | 1987 | 3944034 | ||
BX640839.1 | - | 2 | 2551 |