Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRFAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272053.1 | NP_001258982.1 | 3 | 1820 | ||
NM_001272054.1 | NP_001258983.1 | 3 | 1867 | |||
NM_033296.2 | NP_150638.1 | 2 | 1939 | |||
GenBank mRNA | AA486335.1 | - | 1 | 94 | ||
AK057736.1 | - | 3 | 1293 | |||
AK074496.1 | - | 2 | 1346 | |||
AK123360.1 | - | 3 | 1820 | |||
AK129651.1 | - | 3 | 1224 | |||
AK303974.1 | - | 3 | 1266 | |||
AL539167.3 | - | 1 | 398 | |||
BG255189.1 | - | 1 | 102 | |||
BI858568.1 | - | 1 | 487 |