Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033309.2 | NP_171608.2 | 2 | 2582 | ||
GenBank mRNA | AF171091.1 | - | 1 | 1164 | ||
AY358485.1 | - | 2 | 2514 |