Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005714.1 | NP_005705.1 | 2 | 589 | ||
NM_033347.1 | NP_203133.1 | 2 | 589 | |||
NM_033348.1 | NP_203134.1 | 2 | 589 | |||
NM_033455.1 | NP_258416.1 | 2 | 589 | |||
GenBank mRNA | AF110522.1 | - | 2 | 589 | ||
AF110523.1 | - | 2 | 589 | |||
AF110524.1 | - | 2 | 589 | |||
BC103809.1 | - | 2 | 402 | 40000336 | ||
BC103810.2 | - | 2 | 401 | 40000339 | ||
BC103811.2 | - | 2 | 401 |