Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320199.1 | NP_001307128.1 | 2 | 438 | ||
NM_001320201.1 | NP_001307130.1 | 2 | 388 | |||
NM_033418.3 | NP_219486.1 | 2 | 493 | |||
XM_006711627.3 | XP_006711690.1 | 2 | 946 | |||
GenBank mRNA | AK313909.1 | - | 2 | 413 | ||
AL035369.1 | - | 2 | 407 | |||
AL556604.3 | - | 2 | 483 | |||
AY744366.1 | - | 1 | 129 | |||
BC008679.1 | - | 2 | 388 | 3854146 | ||
BG614785.1 | - | 2 | 365 | |||
CR450330.1 | - | 1 | 129 | |||
HY159746.1 | - | 2 | 493 |