Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291726.1 | NP_001278655.1 | 2 | 310 | ||
NM_001291728.1 | NP_001278657.1 | 2 | 310 | |||
NM_001291730.1 | NP_001278659.1 | 2 | 310 | |||
NM_001291732.1 | NP_001278661.1 | 2 | 310 | |||
NM_001291733.1 | NP_001278662.1 | 2 | 310 | |||
NM_033419.4 | NP_219487.3 | 2 | 310 | |||
XM_011525480.2 | XP_011523782.1 | 2 | 297 | |||
XM_011525481.2 | XP_011523783.1 | 2 | 297 | |||
GenBank mRNA | AB088396.1 | - | 2 | 265 | ||
AK057911.1 | - | 3 | 276 | |||
AK075474.1 | - | 2 | 297 | |||
AK289612.1 | - | 2 | 297 | |||
AK294639.1 | - | 2 | 297 | |||
AK301106.1 | - | 3 | 212 | |||
AK314469.1 | - | 2 | 270 | |||
AY358437.1 | - | 2 | 286 | |||
BC010652.1 | - | 2 | 271 | 3855206 | ||
BG472165.1 | - | 2 | 270 | |||
DC389056.1 | - | 2 | 311 |