Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002944.2 | NP_002935.2 | 3 | 385 | ||
XM_006715548.3 | XP_006715611.1 | 3 | 492 | |||
XM_011536049.2 | XP_011534351.1 | 3 | 493 | |||
XM_011536050.2 | XP_011534352.1 | 3 | 493 | |||
XM_011536051.2 | XP_011534353.1 | 3 | 493 | |||
XM_011536052.2 | XP_011534354.1 | 3 | 493 | |||
XM_011536053.2 | XP_011534355.1 | 3 | 493 | |||
XM_011536054.2 | XP_011534356.1 | 3 | 494 | |||
XM_011536055.2 | XP_011534357.1 | 3 | 493 | |||
XM_011536056.2 | XP_011534358.1 | 3 | 492 | |||
XM_011536057.2 | XP_011534359.1 | 3 | 491 | |||
XM_011536058.2 | XP_011534360.1 | 3 | 491 | |||
XM_017011172.1 | XP_016866661.1 | 3 | 493 | |||
XM_017011173.1 | XP_016866662.1 | 3 | 595 | |||
GenBank mRNA | BC144556.1 | - | 3 | 233 | ||
M34353.1 | - | 3 | 392 |