Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199640.1 | NP_001186569.1 | 3 | 286 | ||
NM_001314044.1 | NP_001300973.1 | 3 | 273 | |||
NM_001314045.1 | NP_001300974.1 | 3 | 281 | |||
NM_001314046.1 | NP_001300975.1 | 3 | 286 | |||
NM_001314047.1 | NP_001300976.1 | 3 | 273 | |||
NM_001314048.1 | NP_001300977.1 | 3 | 286 | |||
NM_033439.3 | NP_254274.1 | 3 | 286 | |||
XM_011518061.2 | XP_011516363.1 | 2 | 221 | |||
XM_017015285.1 | XP_016870774.1 | 5 | 1032 | |||
GenBank mRNA | AB024518.1 | - | 2 | 225 | ||
AK303943.1 | - | 3 | 263 | |||
AK313414.1 | - | 3 | 265 | |||
AY905581.1 | - | 2 | 208 | |||
BC047085.1 | - | 3 | 263 | 5314040 | ||
BQ878541.1 | - | 2 | 247 | |||
CR407619.1 | - | 2 | 208 | |||
DA439244.1 | - | 3 | 281 | |||
DA713547.1 | - | 3 | 273 | |||
DA719342.1 | - | 3 | 271 | |||
DB199599.1 | - | 3 | 295 | |||
DB266668.1 | - | 3 | 265 | |||
DB477860.1 | - | 3 | 286 | |||
HQ641439.1 | - | 2 | 223 |