Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267623.1 | NP_001254552.1 | 2 | 290 | ||
NM_001324236.1 | NP_001311165.1 | 3 | 416 | |||
NM_001324237.1 | NP_001311166.1 | 4 | 321 | |||
NM_001324238.1 | NP_001311167.1 | 3 | 324 | |||
NM_001324240.1 | NP_001311169.1 | 4 | 413 | |||
NM_033453.3 | NP_258412.1 | 4 | 413 | |||
NM_181493.3 | NP_852470.1 | 4 | 362 | |||
NR_052000.1 | 3 | 256 | ||||
NR_052001.2 | 3 | 334 | ||||
NR_052002.1 | 3 | 348 | ||||
XM_006723564.3 | XP_006723627.1 | 4 | 696 | |||
XM_006723565.3 | XP_006723628.1 | 2 | 573 | |||
XM_011529234.2 | XP_011527536.1 | 4 | 696 | |||
GenBank mRNA | AB062127.1 | - | 4 | 282 | ||
AF026816.2 | - | 4 | 272 | |||
AF063607.1 | - | 4 | 297 | |||
AF219116.1 | - | 4 | 325 | |||
AF334680.1 | - | 3 | 205 | |||
AK309165.1 | - | 3 | 275 | |||
AV694278.1 | - | 3 | 206 | |||
BC010138.2 | - | 4 | 257 | 4124624 | ||
BE275997.1 | - | 3 | 291 | |||
BF969192.1 | - | 3 | 256 | |||
BI115811.1 | - | 2 | 126 | |||
BM560812.1 | - | 413 | ||||
BM703982.1 | - | 3 | 233 | |||
BQ049090.1 | - | 3 | 195 | |||
EF199841.1 | - | 4 | 260 | |||
EF213026.1 | - | 4 | 209 | |||
KU178148.1 | - | 4 | 221 | |||
KU178149.1 | - | 2 | 98 |