Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC37B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321350.1 | NP_001308279.1 | 4 | 510 | ||
NM_052888.2 | NP_443120.2 | 1 | 316 | |||
GenBank mRNA | AJ314647.1 | - | 1 | 302 | ||
AK299772.1 | - | 4 | 364 | |||
AK302740.1 | - | 1 | 447 | |||
AK310287.1 | - | 1 | 344 | |||
AY386261.1 | - | 2 | 354 | |||
BC117373.1 | - | 1 | 355 | 40125924 | ||
BC143578.1 | - | 1 | 355 | |||
BC143581.1 | - | 1 | 355 | 9052092 | ||
BF979626.1 | - | 1 | 348 | |||
DB475546.2 | - | 2 | 320 |