Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052886.2 | NP_443118.1 | 4 | 2302 | ||
XM_011516807.2 | XP_011515109.1 | 3 | 2086 | |||
GenBank mRNA | AK172820.1 | - | 4 | 2211 | ||
AL117612.1 | - | 1 | 1593 | |||
AY007723.1 | - | 4 | 2279 |