Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197327.1 | NP_001184256.1 | 6 | 630 | ||
NM_001318322.1 | NP_001305251.1 | 5 | 532 | |||
NM_052970.4 | NP_443202.3 | 6 | 630 | |||
XM_011529151.2 | XP_011527453.1 | 3 | 328 | |||
XM_017027632.1 | XP_016883121.1 | 6 | 545 | |||
XM_017027633.1 | XP_016883122.1 | 6 | 529 | |||
XM_017027634.1 | XP_016883123.1 | 6 | 529 | |||
XM_017027635.1 | XP_016883124.1 | 6 | 573 | |||
GenBank mRNA | AK056712.1 | - | 6 | 567 | ||
AK297120.1 | - | 4 | 423 | |||
AK301294.1 | - | 5 | 498 | |||
BC090857.1 | - | 5 | 491 | |||
BC110881.1 | - | 6 | 579 | 5725594 | ||
BC117284.1 | - | 6 | 528 | 40125835 | ||
BC126263.1 | - | 6 | 528 | 8991979 | ||
BC143932.1 | - | 6 | 528 | 9052450 | ||
CB990660.1 | - | 6 | 661 |